Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XAGE5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565989 Supplier Invitrogen™ Supplier No. PA565989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRRYRPRRCLR LAQLVGPMLE PSVPEPQ Highest antigen sequence identity to the following orthologs - mouse 46%, rat 39%.

This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. The protein encoded by this gene shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XAGE5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XAGE5
Gene Accession No. Q8WWM1
Gene Alias Cancer/testis antigen 12.5; cancer/testis antigen family 12, member 5; CT12.5; G antigen family D member 5; G antigen, family D, 5; GAGED5; protein XAGE-5; X antigen family member 5; X antigen family, member 5; XAGE5; XAGE-5
Gene Symbols XAGE5
Host Species Rabbit
Immunogen Recombinant Human XAGE5. Recombinant protein control fragment (Product #RP-106672).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170627
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.