Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Xanomeline oxalate
GREENER_CHOICE

Catalog No. 356910R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-691-0R 10 mg
35-695-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-691-0R Supplier Tocris Bioscience™ Supplier No. 3569/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Functionally selective M1 agonist

Xanomeline oxalate is a functionally selective muscarinic M1 receptor agonist (EC50 values are 0.3, 5, 42, 52 and 92.5 nM at M1, M3, M5, M4 and M2 receptors respectively). Displays a complex pharmacological profile: reversible and wash-resistant binding, resulting in full agonist activity at M1; delayed wash-resistant partial agonist activity at M2; and delayed wash-resistant full agonist activity at M4. Exhibits antipsychotic activity, and improves cognitive deficits and behavioral disturbances in Alzheimer's disease and schizophrenia.

Specifications

Quantity 10 mg
Product Type Xanomeline oxalate
Purity >98%
Molecular Weight (g/mol) 371.46
Inhibitors Xanomeline oxalate

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.