Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
69-89-6
C5H4N4O2
152.11
MFCD00078453
LRFVTYWOQMYALW-UHFFFAOYSA-N
xanthine,2,6-dihydroxypurine,xanthin,2,6-dioxopurine,pseudoxanthine,1h-purine-2,6 3h,7h-dione,isoxanthine,xanthic oxide,1h-purine-2,6-diol,9h-purine-2,6-diol
1188
CHEBI:17712
3,7-dihydropurine-2,6-dione
C1=NC2=C(N1)C(=O)NC(=O)N2
69-89-6
C5H4N4O2
152.11
MFCD00078453
LRFVTYWOQMYALW-UHFFFAOYSA-N
xanthine,2,6-dihydroxypurine,xanthin,2,6-dioxopurine,pseudoxanthine,1h-purine-2,6 3h,7h-dione,isoxanthine,xanthic oxide,1h-purine-2,6-diol,9h-purine-2,6-diol
1188
CHEBI:17712
3,7-dihydropurine-2,6-dione
C1=NC2=C(N1)C(=O)NC(=O)N2
Sulfated Ash 0.2% max. (800°C)
Quantity 25g
Loss on Drying 1% max. (105°C, 5 hrs)
Packaging Glass bottle
Melting Point >300.0°C
Color White to Yellow
Infrared Spectrum Authentic
Appearance (c=1, 1M NaOH) clear colorless to light yellowsoln
Spectrophotometry 98 to 102% (lambda max. 278nm, pH 10), molar absorptivity 9300 +/- 2%, on dry basis
Assay Percent Range 98%
Beilstein 26, 447
Merck Index 15, 10257
Solubility Information Solubility in water: 1g/14.5l (16°C). Other solubilities: soluble in potassium hydroxide and mineral acids,practically insoluble in most common organic,solvents
Formula Weight 152.11
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Xanthine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning