Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthine, 99%

Catalog No. p-7081206
Click to view available options
Quantity:
25 g
100 g

CAS: 69-89-6 | C5H4N4O2 | 152.113 g/mol

Xanthine and xanthine oxidase system can be used to produce superoxide radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.

Solubility
Soluble in water(0.067g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-89-6
Molecular Formula C5H4N4O2
Molecular Weight (g/mol) 152.113
MDL Number MFCD00078453
InChI Key LRFVTYWOQMYALW-UHFFFAOYSA-N
Synonym xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol
PubChem CID 1188
ChEBI CHEBI:17712
IUPAC Name 3,7-dihydropurine-2,6-dione
SMILES C1=NC2=C(N1)C(=O)NC(=O)N2

Specifications

Melting Point >300°C
Odor Odorless
Quantity 25 g
Beilstein 8733
Merck Index 14,10059
Solubility Information Soluble in water(0.067g/L).
Formula Weight 152.11
Percent Purity 99%
Chemical Name or Material Xanthine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-718-6
RTECSNumber ZD7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.