Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Xanthine Oxidase Antibody (8A9), Novus Biologicals™

Rabbit Monoclonal Antibody
$305.98 - $479.73
Specifications
| Antigen | Xanthine Oxidase |
|---|---|
| Clone | 8A9 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
Description
Xanthine Oxidase Monoclonal antibody specifically detects Xanthine Oxidase in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| Xanthine Oxidase | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.17.1.4, EC 1.7.2.2, xanthene dehydrogenase, xanthine dehydrogenase, xanthine dehydrogenase/oxidase, xanthine oxidase, xanthine oxidoreductase, XDHA, XO, XOR | |
| A synthesized peptide derived from Human Xanthine Oxidase [UniProt P47989] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8A9 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 7498 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title