Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Xanthine Oxidase Mouse anti-Human, Clone: 1C8, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275716
Description
Xanthine Oxidase Monoclonal specifically detects Xanthine Oxidase in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Xanthine Oxidase | |
Monoclonal | |
2 mg/mL | |
Flow Cytometry 1:10-1:1000, ELISA 1:5000-1:10000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
EC 1.17.1.4, EC 1.7.2.2, xanthene dehydrogenase, xanthine dehydrogenase, xanthine dehydrogenase/oxidase, xanthine oxidase, xanthine oxidoreductase, XDHA, XO, XOR | |
Mouse | |
146 kDa | |
100 μL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1C8 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide | |
XDH | |
Synthetic peptide within Human Xanthine Oxidase aa 1,082-1,131/ 1,333. (SwissProt: P47989 Human) | |
Protein G purified | |
Signal Transduction | |
7498.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction