Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Xanthohumol
SDP

Catalog No. 468610
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-861-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-861-0 Supplier Tocris Bioscience™ Supplier No. 4686/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

p97 ATPase (VCP) inhibitor

p97 ATPase (VCP) inhibitor. Binds to the N domain of p97, impairing autophagosome function and maturation. Inhibits proliferation and induces apoptosis in a variety of human cancer cell lines. Also inhibits DGAT in rat liver microsomes (IC50 = 50.3 μM).

Chemical Identifiers

CAS 6754-58-1
Molecular Formula C21H22O5

Specifications

Quantity 10 mg
Formula Weight 354.4
Purity >98%
Molecular Weight (g/mol) 354.402
Chemical Name or Material Xanthohumol
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.