Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthone, 99%

Catalog No. p-7022175
Click to view available options
Quantity:
25 g
100 g

CAS: 90-47-1 | C13H8O2 | 196.205 g/mol

Xanthone is used predominantly as oxygenated polycyclic aromatic compounds, and as a vasorelaxant. It can be used as a fluorescent agent, as well as an intermediate in organic synthesis and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xanthone is used predominantly as oxygenated polycyclic aromatic compounds, and as a vasorelaxant. It can be used as a fluorescent agent, as well as an intermediate in organic synthesis and chemical research.

Solubility
It is Soluble in water (partly), chloroform, hot Toluene, ether (slightly), and hot alcohol.

Notes
Stable under normal conditions. Incompatible to strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-47-1
Molecular Formula C13H8O2
Molecular Weight (g/mol) 196.205
MDL Number MFCD00005060
InChI Key JNELGWHKGNBSMD-UHFFFAOYSA-N
Synonym xanthone, 9h-xanthen-9-one, 9-xanthenone, benzophenone oxide, 9-oxoxanthene, genicide, xanthenone, 9-xanthone, diphenylene ketone oxide, dibenzo-gamma-pyrone
PubChem CID 7020
ChEBI CHEBI:37647
IUPAC Name xanthen-9-one
SMILES C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3O2

Specifications

Melting Point 174°C to 178°C
Boiling Point 350°C to 351°C
Quantity 25 g
Beilstein 140443
Merck Index 14,10062
Solubility Information It is Soluble in water (partly),chloroform,hot Toluene,ether (slightly),and hot alcohol.
Formula Weight 196.21
Percent Purity 99%
Chemical Name or Material Xanthone
EINECSNumber 201-997-7
RTECSNumber ZD5711000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.