Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthydrol, 98+%

Catalog No. aaa1347606
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 90-46-0 | C13H10O2 | 198.221 g/mol

Useful in tests for urea

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-46-0
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00005057
InChI Key JFRMYMMIJXLMBB-UHFFFAOYSA-N
Synonym 9-hydroxyxanthene, xanthydrol, xanthanol, 9-xanthydrol, xanthen-9-ol, xanthene, hydroxy, 9-xanthenol, ccris 1640, unii-7131m69ikf, xanthrol
PubChem CID 72861
IUPAC Name 9H-xanthen-9-ol
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)O

Specifications

Melting Point 122°C to 126°C
Quantity 5 g
Beilstein 10395
Sensitivity Light sensitive
Formula Weight 198.22
Percent Purity ≥98%
Chemical Name or Material Xanthydrol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-996-1
RTECSNumber ZD5710000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.