Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ XAP 044
GREENER_CHOICESDP

Catalog No. 524850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-485-0 50 mg
52-481-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-485-0 Supplier Tocris Bioscience™ Supplier No. 5248/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective mGlu7 antagonist

XAP 044 is a potent and selective mGlu7 antagonist (IC50 = 88 nM); inhibits lateral amygdala long term potentiation (LTP) in brain slices from wild type mice. Displays no effects on the LTP of mGlu7 deficient mice. Exhibits novel activity by binding to the mGlu7 Venus flytrap domain (VFTD). Exhibits anti-anxiety effects in rodent models. Brain penetrant.

Specifications

Quantity 50 mg
Product Type XAP 044
Purity >98%
Molecular Weight (g/mol) 380.13
Inhibitors XAP 044

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.