Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ XAV 939
GREENER_CHOICESDP

Catalog No. 374850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-485-0 50 mg
37-481-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-485-0 Supplier R&D Systems™ Supplier No. 3748/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent tankyrase inhibitor

XAV 939 is a potent tankyrase (TNKS) inhibitor (IC50 values are 4 and 11 nM for TNKS2 and TNKS1 respectively). Antagonizes Wnt signaling via stimulation of β-catenin degradation and stabilization of axin. XAV 939 inhibits proliferation of the β-catenin-dependent colon carcinoma cell line DLD-1. Promotes cardiomyocyte differentiation in mesoderm progenitor cells. Also used in neuronal differentiation protocols.

XAV 939 synthesized to cGMP guidelines also available.

For more information about how XAV 939 may be used, see our protocol: Accelerated Induction of Cortical Neurons from hiPSCs

Chemical Identifiers

CAS 284028-89-3
Molecular Formula C14H11F3N2OS

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 312.31
Purity >98%
Molecular Weight (g/mol) 312.31
Chemical Name or Material XAV 939
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.