Learn More
Description
Research use only XAV 939 also available.
Specifications
Specifications
| Chemical Name or Material | 3,5,7,8-Tetrahydro-2-[4-(trifluoromethyl)phenyl]-4H-thiopyrano[4,3-d]pyrimidin-4-one |
| CAS | 284028-89-3 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Tankyrase Inhibitors |
| Molecular Formula | C14H11F3N2OS |
| Purity | 99% |
| Regulatory Status | GMP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
