Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XBP1 Antibody (3H1G4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP175514PCP
Description
XBP1 Monoclonal antibody specifically detects XBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
XBP1 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunocytochemistry | |
3H1G4 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5 | |
Antibody was raised against recombinant protein corresponding to amino acids 2 to 160 of human XBP-1. | |
0.1 mL | |
Cancer, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
7494 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction