Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XBP1 Antibody (525904), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4257I
Description
XBP1 Monoclonal antibody specifically detects XBP1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
XBP1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
525904 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5 | |
E. coli-derived recombinant human XBP1S, Ser188-Leu287, Accession # NP_001073007 | |
0.1 mL | |
Cancer, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
7494 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction