Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XBP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP220917
Description
XBP1 Polyclonal specifically detects XBP1 in Human, Mouse, Porcine samples. It is validated for Western Blot, Knockout Validated, Knockdown Validated.Specifications
XBP1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5 | |
Rabbit | |
29 kDa | |
0.1 mL | |
Cancer, Lipid and Metabolism, Unfolded Protein Response | |
7494 | |
Human, Mouse, Pig | |
IgG |
Western Blot, Immunoassay, KnockDown | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
P17861 | |
XBP1 | |
Recombinant protein encompassing a sequence within the center region of human XBP1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: cow (89%), Mouse (82%), Rat (86%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction