Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595907 Supplier Invitrogen™ Supplier No. PA595907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T, NCI-H460, Mouse eye, Rat liver, Rat eye; Cellular Location: Cytoplasm, Endoplasmic reticulum membrane, Endoplasmic reticulum, Membrane, Nucleus, Peripheral membrane protein, Single-pass type II membrane protein Immunogen sequence: NVVVKIEEAP LSPSENDHPE FIVSVKEEPV EDDLVPELGI SNLLSSSHCP KPSSCLLDAY SDCGYGGSLS PFSDMSSLLG VNHSWEDTFA NELFPQLISV.

XBP1 encodes a transcription factor that regulates MHC class II genes by binding to a promoter element referred to as an X box. This gene product is a bZIP protein, which was also identified as a cellular transcription factor that binds to an enhancer in the promoter of the T cell leukemia virus type 1 promoter. It may increase expression of viral proteins by acting as the DNA binding partner of a viral transactivator. It has been found that upon accumulation of unfolded proteins in the endoplasmic reticulum (ER), the mRNA of this gene is processed to an active form by an unconventional splicing mechanism that is mediated by the endonuclease inositol-requiring enzyme 1 (IRE1). The resulting loss of 26 nt from the spliced mRNA causes a frame-shift and an isoform XBP1(S), which is the functionally active transcription factor. The isoform encoded by the unspliced mRNA, XBP1(U), is constitutively expressed, and thought to function as a negative feedback regulator of XBP1(S), which shuts off transcription of target genes during the recovery phase of ER stress. A pseudogene of XBP1 has been identified and localized to chromosome 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XBP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Xbp1
Gene Accession No. O35426, P17861, Q9R1S4
Gene Alias D11Ertd39e; Hepatocarcinogenesis-related transcription factor; HTF; OTTHUMP00000028682; Tax-responsive element-binding protein 5; tax-responsive element-binding protein 5 homolog; TREB5; TREB-5; X-box binding protein 1; X-box-binding protein 1; X-box-binding protein 1, cytoplasmic form; X-box-binding protein 1, luminal form; XBP1; XBP-1; XBP2
Gene Symbols Xbp1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 277-376 of human XBP1 (NP_001073007.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22433, 289754, 7494
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.