Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation XBridge BEH HILIC XP Column

Catalog No. 50465928 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Quantity:
1/Ea.
3/Pk.
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Quantity
50-465-928 150 mm 3 mm 3/Pk.
50-179-499 50 mm 2.1 mm 1/Ea.
50-179-237 30 mm 2.1 mm 3/Pk.
50-179-238 50 mm 2.1 mm 3/Pk.
50-179-239 75 mm 2.1 mm 3/Pk.
50-179-240 100 mm 2.1 mm 3/Pk.
50-179-241 30 mm 3 mm 3/Pk.
50-179-242 50 mm 3 mm 3/Pk.
50-179-243 75 mm 3 mm 3/Pk.
50-179-244 100 mm 3 mm 3/Pk.
50-179-498 30 mm 2.1 mm 1/Ea.
50-179-500 75 mm 2.1 mm 1/Ea.
50-179-501 100 mm 2.1 mm 1/Ea.
50-179-502 30 mm 3 mm 1/Ea.
50-179-503 50 mm 3 mm 1/Ea.
50-179-504 75 mm 3 mm 1/Ea.
50-179-505 100 mm 3 mm 1/Ea.
50-201-530 30 mm 4.6 mm 1/Ea.
50-201-531 50 mm 4.6 mm 1/Ea.
50-201-532 75 mm 4.6 mm 1/Ea.
50-201-533 100 mm 4.6 mm 1/Ea.
50-465-927 150 mm 2.1 mm 3/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. 50-465-928 Supplier Waters Corporation Supplier No. 176002888
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds.

  • These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica based HILIC phases, the BEH particle's chemical stability results in long column lifetimes.
  • Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.

Specifications

Diameter (Metric) 3 mm
Length (Metric) 150 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 2.5 μm
Pore Size 130 Å
Quantity 3/Pk.
USP Type L3
Endcapped No
Product Type eXtended Performance (XP) Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.