Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation XBridge BEH130 C18 Column

Catalog No. 50781923 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4.6 mm
Particle Size:
3.5 μm
5 μm
10 μm
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size
50-781-923 250 mm 4.6 mm 10 μm
50-781-843 50 mm 2.1 mm 3.5 μm
50-781-857 250 mm 2.1 mm 5 μm
50-781-840 50 mm 1 mm 3.5 μm
50-781-841 100 mm 1 mm 3.5 μm
50-781-842 150 mm 1 mm 3.5 μm
50-781-844 100 mm 2.1 mm 3.5 μm
50-781-845 150 mm 2.1 mm 3.5 μm
50-781-846 250 mm 2.1 mm 3.5 μm
50-781-847 50 mm 4.6 mm 3.5 μm
50-781-848 100 mm 4.6 mm 3.5 μm
50-781-849 150 mm 4.6 mm 3.5 μm
50-781-850 250 mm 4.6 mm 3.5 μm
50-781-851 50 mm 1 mm 5 μm
50-781-852 100 mm 1 mm 5 μm
50-781-853 150 mm 1 mm 5 μm
50-781-854 50 mm 2.1 mm 5 μm
50-781-855 100 mm 2.1 mm 5 μm
50-781-856 150 mm 2.1 mm 5 μm
50-781-858 50 mm 4.6 mm 5 μm
50-781-859 100 mm 4.6 mm 5 μm
50-781-860 150 mm 4.6 mm 5 μm
50-781-861 250 mm 4.6 mm 5 μm
50-781-920 50 mm 4.6 mm 10 μm
50-781-921 100 mm 4.6 mm 10 μm
50-781-922 150 mm 4.6 mm 10 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Catalog No. 50-781-923 Supplier Waters Corporation Supplier No. 186003651
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations.

  • The small-pore (130 Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 10 μm
Pore Size 130 Å
Quantity 1/Ea.
USP Type L1
Endcapped Yes
For Use With (Application) Peptides
Product Type Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.