Learn More
Waters Corporation XBridge BEH300 C18 Method Validation Kit
Shop All Waters Chromatography Solutions Products
Description
- The reproducibility of a chromatographic column has a critical impact on the long-term reliability and robustness of an analytical method, and its reproducibility lies completely outside the user's control.
- With exceptional batch-to-batch and column-to-column reproducibility, Waters well-established particle and column manufacturing process control provides confidence in the long-term reliability of your analytical method.
- Each batch of selected XBridge BEH300 C18 chemistry is specifically QC tested with a mixture of carefully selected peptide standards to help ensure consistent performance.
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 100 mm |
| Max. Pressure | 6000 psi (415 Bar) |
| Particle Shape | Spherical |
| Particle Size | 5 μm |
| Pore Size | 130 Å |
| Quantity | 3/Pk. |
| USP Type | L1 |
| Endcapped | No |
| For Use With (Application) | Peptides |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.