Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ XCC
GREENER_CHOICESDP

Catalog No. 422010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-201-0 10 mg
42-205-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-201-0 Supplier Tocris Bioscience™ Supplier No. 4220/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Adenosine receptor antagonist

Adenosine receptor antagonist (Ki values are 42, 68 and 1130 nM for A1, A2B and A2A receptors).

Chemical Identifiers

CAS 96865-83-7
Molecular Formula C19H22N4O5

Specifications

Quantity 10 mg
Formula Weight 386.4
Purity >98%
Molecular Weight (g/mol) 386.408
Chemical Name or Material XCC
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.