Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XCL1/Lymphotactin Antibody (001), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290947AF532
Description
XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Cynomolgus Monkey samples. It is validated for ELISA.Specifications
XCL1/Lymphotactin | |
Monoclonal | |
Alexa Fluor 532 | |
ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus XCL1/Lymphotactin (; Met1-Gly114). | |
0.1 mL | |
Biologically Active Proteins, Immunology, Innate Immunity | |
6375 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Cynomolgus Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction