Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XCL1/Lymphotactin Antibody (MM0604-4G24) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018987 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018987 0.025 mg
NBP212004 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018987 Supplier Novus Biologicals Supplier No. NBP2120040.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Human samples. It is validated for Western Blot.

Specifications

Antigen XCL1/Lymphotactin
Applications Western Blot
Classification Monoclonal
Clone MM0604-4G24
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. P47992
Gene Alias ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin)
Gene Symbols XCL1
Host Species Mouse
Immunogen Human Lymphotactin
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chemokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 6375
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.