Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XCL1/XCL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575840
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575840 Supplier Invitrogen™ Supplier No. PA575840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Lymphotactin, otherwise known as XCL1, is a member of the C-chemokine family that uniquely acts as a chemoattractant for lymphocytes, but not monocytes or neutrophils, suggesting an important regulatory role in lymphocyte trafficking and inflammation. Lymphotactin, shown to signal through chemokine XC receptor 1 (XCR1), is detected at the highest level in the spleen and at low levels in the lungs, small intestine and colon, and is abundantly and selectively expressed by activated T cells, in particular CD8+ T cells, and is also produced by NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XCL1/XCL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene XCL1
Gene Accession No. P47992, P47993, P51672, Q9UBD3
Gene Alias AI661682; ATAC; C motif chemokine 1; c motif chemokine 2; chemokine (C motif) ligand 1; chemokine (C motif) ligand 2; cytokine SCM-1; Cytokine SCM-1 beta; Lptn; LTN; Lymphotactin; lymphotaxin; SCM1; SCM-1; SCM1A; SCM-1a; SCM-1-alpha; SCM1B; SCM-1b; Scyc1; SCYC2; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); small inducible cytokine subfamily C, member 2; small-inducible cytokine C1; XC chemokine ligand 1; XC chemokine ligand 2; X-C motif chemokine ligand 1; X-C motif chemokine ligand 2; Xcl1; XCL2
Gene Symbols XCL1, XCL2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human XCL1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16963, 171371, 6375, 6846
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.