Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XCL2 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306178R
Description
XCL2 Polyclonal specifically detects XCL2 in Human samples. It is validated for ELISA.Specifications
XCL2 | |
Polyclonal | |
ELISA | |
C motif chemokine 2, chemokine (C motif) ligand 2, cytokine SCM-1 beta, SCM-1b, SCM1B, SCYC2XC chemokine ligand 2, small inducible cytokine subfamily C, member 2 | |
Produced in rabbits immunized with purified, recombinant Human XCL2 (Q9UBD3; Met1-Gly114). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
6846 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction