Learn More
Invitrogen™ XCL2 Monoclonal Antibody (06)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human XCL2.
Specifications
Specifications
| Antigen | XCL2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 06 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | XCL2 |
| Gene Accession No. | Q9UBD3 |
| Gene Alias | C motif chemokine 2; chemokine (C motif) ligand 2; cytokine SCM-1 beta; SCM1B; SCM-1b; SCYC2; small inducible cytokine subfamily C, member 2; XC chemokine ligand 2; X-C motif chemokine ligand 2; XCL2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.