Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XCL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575915
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Chemotactic activity for lymphocytes but not for monocytes or neutrophils. [UniProt].
Specifications
XCL2 | |
Polyclonal | |
Unconjugated | |
XCL2 | |
C motif chemokine 2; chemokine (C motif) ligand 2; cytokine SCM-1 beta; SCM1B; SCM-1b; SCYC2; small inducible cytokine subfamily C, member 2; XC chemokine ligand 2; X-C motif chemokine ligand 2; XCL2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6846 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBD3 | |
XCL2 | |
Synthetic peptide corresponding to residues in Human XCL2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction