Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human XCR1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB8571V100UG
This item is not returnable.
View return policy
Description
XCR1/CCXCR1 Monoclonal specifically detects XCR1/CCXCR1 in Human samples. It is validated for Flow Cytometry.Specifications
XCR1/CCXCR1 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CCXCR1, CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor | |
E. coli-derived recombinant human XCR1 Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267 Accession # P46094 | |
RUO | |
2829 | |
Human | |
IgG |
Flow Cytometry | |
1097A | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P46094 | |
Rabbit | |
100 μg | |
Primary | |
Detects human XCR1 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction