Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
xCT Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10059735
Description
xCT Polyclonal specifically detects xCT in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
xCT | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, cystine/glutamate transporter, Member 11 (Slc7a11), Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCTCCBR1 | |
Goat | |
55 kDa | |
0.1 mg | |
Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
23657 | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), ELISA, Immunofluorescence | |
0.5 mg/mL | |
Western Blot 0.03 - 0.3 ug/mL, Flow Cytometry 10 ug/mL, Immunohistochemistry-Paraffin 3.75 ug/mL, Peptide ELISA Detection limit 1:128000, Immunofluorescence 10 ug/mL | |
NP_055146.1 | |
SLC7A11 | |
Peptide with sequence C-KGQTQNFKDAFSGRD corresponding to internal region according to NP_055146.1. | |
Affinity Purified | |
RUO | |
Primary | |
Expected from sequence similarity: Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction