Learn More
Invitrogen™ XDH Recombinant Rabbit Monoclonal Antibody (0M8E7)

Description
Immunogen sequence: TPLDPTQEPI FPPELLRLKD TPRKQLRFEG ERVTWIQAST LKELLDLKAQ HPDAKLVVGN TEIGIEMKFK NMLFPMIVCP AWIPELNSVE HG.
Specifications
Specifications
| Antigen | XDH |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 0M8E7 |
| Concentration | 0.37 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | XDH |
| Gene Accession No. | P22985, P47989, Q00519 |
| Gene Alias | Unknown (protein for MGC:127098); xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; xanthine oxidase; Xanthine oxidoreductase; XD; Xdh; xdh antibody; XDHA; xdha antibody; XO; XOR; xor antibody; Xox1; Xox-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.