Learn More
XDH Mouse anti-Human, Clone: OTI1C10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812777

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
XDH belongs to the group of molybdenum-containing hydroxylases involved in the oxidative metabolism of purines. The enzyme is a homodimer. This protein can be converted to xanthine oxidase by reversible sulfhydryl oxidation or by irreversible proteolytic modification. Defects in xanthine dehydrogenase cause xanthinuria, may contribute to adult respiratory stress syndrome, and may potentiate influenza infection through an oxygen metabolite-dependent mechanism.Specifications
XDH | |
Monoclonal | |
Unconjugated | |
XDH | |
XAN1, XO, XOR | |
Mouse | |
Affinity Chromatography | |
RUO | |
7498 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1C10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P47989 | |
XDH | |
Human recombit protein fragment corresponding to amino acids 1-280 of human XDH produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.