Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526285
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526285 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526285 Supplier Invitrogen™ Supplier No. PA526285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XDH Polyclonal Antibody for Western Blot, IHC (P), Flow

XDH belongs to the group of molybdenum-containing hydroxylases involved in the oxidative metabolism of purines. The enzyme is a homodimer. This protein can be converted to xanthine oxidase by reversible sulfhydryl oxidation or by irreversible proteolytic modification. Defects in xanthine dehydrogenase cause xanthinuria, may contribute to adult respiratory stress syndrome, and may potentiate influenza infection through an oxygen metabolite-dependent mechanism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XDH
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% ProClin 300; pH 7.4
Gene XDH
Gene Accession No. P47989, Q00519
Gene Alias Unknown (protein for MGC:127098); xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; xanthine oxidase; Xanthine oxidoreductase; XD; Xdh; xdh antibody; XDHA; xdha antibody; XO; XOR; xor antibody; Xox1; Xox-1
Gene Symbols XDH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 206-234 amino acids from the N-terminal region of human XDH.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22436, 7498
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.