Learn More
Invitrogen™ XDH Polyclonal Antibody

Description
Immunogen sequence: GFVCFISADDV PGSNITGICN DETVFAKDKV TCVGHIIGAV VADTPEHTQR AAQGVKITYE ELPAIITIED AIKNNSFYGP ELKIEKGDLK K Highest antigen sequence identity to the following orthologs - mouse 87%, rat 84%.
Specifications
Specifications
| Antigen | XDH |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | XDH |
| Gene Accession No. | P47989 |
| Gene Alias | Unknown (protein for MGC:127098); xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; xanthine oxidase; Xanthine oxidoreductase; XD; Xdh; xdh antibody; XDHA; xdha antibody; XO; XOR; xor antibody; Xox1; Xox-1 |
| Gene Symbols | XDH |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.