Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567586 Supplier Invitrogen™ Supplier No. PA567586
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XDH Polyclonal Antibody for IHC (P)

XDH belongs to the group of molybdenum-containing hydroxylases involved in the oxidative metabolism of purines. The enzyme is a homodimer. This protein can be converted to xanthine oxidase by reversible sulfhydryl oxidation or by irreversible proteolytic modification. Defects in xanthine dehydrogenase cause xanthinuria, may contribute to adult respiratory stress syndrome, and may potentiate influenza infection through an oxygen metabolite-dependent mechanism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XDH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene XDH
Gene Accession No. P47989
Gene Alias Unknown (protein for MGC:127098); xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; xanthine oxidase; Xanthine oxidoreductase; XD; Xdh; xdh antibody; XDHA; xdha antibody; XO; XOR; xor antibody; Xox1; Xox-1
Gene Symbols XDH
Host Species Rabbit
Immunogen Synthesized peptide derived from human XDH.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7498
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.