Learn More
Invitrogen™ XDH Polyclonal Antibody

Description
Positive Samples: Mouse liver; Cellular Location: Cytoplasm, Peroxisome Immunogen sequence: ETSTNTVPNT SPTAASVSA.
Specifications
Specifications
| Antigen | XDH |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.63 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | XDH |
| Gene Accession No. | P22985, P47989, Q00519 |
| Gene Alias | Unknown (protein for MGC:127098); xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; xanthine oxidase; Xanthine oxidoreductase; XD; Xdh; xdh antibody; XDHA; xdha antibody; XO; XOR; xor antibody; Xox1; Xox-1 |
| Gene Symbols | XDH |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.