Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XDH Recombinant Rabbit Monoclonal Antibody (4B7L4)
GREENER_CHOICE

Catalog No. FEMA551417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551417 Supplier Invitrogen™ Supplier No. MA551417
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: CIIPTKFGIS FTVPFLNQAG ALLHVYTDGS VLLTHGGTEM GQGLHTKMVQ VASRALKIPT SKIYISETST NTVPNTSPTA ASVSADLNGQ AVYAACQTIL K.

XDH belongs to the group of molybdenum-containing hydroxylases involved in the oxidative metabolism of purines. The enzyme is a homodimer. This protein can be converted to xanthine oxidase by reversible sulfhydryl oxidation or by irreversible proteolytic modification. Defects in xanthine dehydrogenase cause xanthinuria, may contribute to adult respiratory stress syndrome, and may potentiate influenza infection through an oxygen metabolite-dependent mechanism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XDH
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4B7L4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene XDH
Gene Accession No. P22985, Q00519
Gene Alias Unknown (protein for MGC:127098); xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; xanthine oxidase; Xanthine oxidoreductase; XD; Xdh; xdh antibody; XDHA; xdha antibody; XO; XOR; xor antibody; Xox1; Xox-1
Gene Symbols XDH
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1000-1100 of human Xanthine Oxidase (XDH) (NP_000370.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22436, 497811
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.