Learn More
Agilent Technologies™ XF HS Mini FluxPak PDL Plates Kit
Catalog No. 103724100
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
10-372-4100 1 Kit
1 options
Catalog No. 10-372-4100 Supplier Agilent Technologies™ Supplier No. 103724100
Add to Cart
Add to Cart
XF HS Mini FluxPak PDL Plates Kit contains 12 XFp sensor cartridges, 12 HS PDL Miniplates, and one 100 mL bottle of calibrant.
  • Available in a wide range of configurations to accommodate 2D monolayers, 3D spheroids, suspension cells, islets, and isolated mitochondria
  • Solid-state sensor probes containing polymer embedded fluorophores detect oxygen and proton levels in cell culture media
  • Patented integrated injection ports for adding compounds, stimulators, inhibitors, substrates, and beads conjugated with antigens
  • Patented transient microchamber provides increased analytical sensitivity and signal to noise of OCR and ECAR (PER, gPER) readings when used in an XF Analyzer
  • Sensors are spotted onto the cartridge using a multi-step process that includes manual and machine vision inspection
  • Conforms to ANSI/SLAS Microplate Standards for footprint dimensions and well positions
  • Pre-coated Poly-D-Lysine cell culture plates, available in 96-well and 8-well formats, are available for use with XF assays using suspension cell, providing convenience and consistent assay result
  • PDL cell culture plates are manufactured in an aseptic facility
  • XF Pro M cell culture plates are now available for use with XF Pro analyzer to minimize assay edge effect and improve intra-plate consistency
  • Designed specifically for use with Seahorse XF Analyzers
Specifications
For Use With (Application) Cell Function
For Use With (Equipment) Other Fluorescence Detection Systems
Product Type Calibration Kit
Quantity 1 Kit