Learn More
Agilent Technologies™ XF HS Mini FluxPak
Catalog No. 103723100
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
10-372-3100 1 Kit
1 options
Catalog No. 10-372-3100 Supplier Agilent Technologies™ Supplier No. 103723100
Add to Cart
Add to Cart
XF HS Mini FluxPak contains 12 XFp sensor cartridges, 12 HS Miniplates, and one 100 mL bottle of calibrant.
  • Available in a wide range of configurations to accommodate 2D monolayers, 3D spheroids, suspension cells, islets, and isolated mitochondria
  • Solid-state sensor probes containing polymer embedded fluorophores detect oxygen and proton levels in cell culture media
  • Patented integrated injection ports for adding compounds, stimulators, inhibitors, substrates, and beads conjugated with antigens
  • Patented transient microchamber provides increased analytical sensitivity and signal to noise of OCR and ECAR (PER, gPER) readings when used in an XF Analyzer
  • Sensors are spotted onto the cartridge using a multi-step process that includes manual and machine vision inspection
  • Conforms to ANSI/SLAS Microplate Standards for footprint dimensions and well positions
  • Pre-coated Poly-D-Lysine cell culture plates, available in 96-well and 8-well formats, are available for use with XF assays using suspension cell, providing convenience and consistent assay result
  • PDL cell culture plates are manufactured in an aseptic facility
  • XF Pro M cell culture plates are now available for use with XF Pro analyzer to minimize assay edge effect and improve intra-plate consistency
  • Designed specifically for use with Seahorse XF Analyzers
Specifications
For Use With (Application) Cell Function
For Use With (Equipment) Other Fluorescence Detection Systems
Product Type Calibration Kit
Quantity 1 Kit