Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XIAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$243.50 - $366.00
Specifications
Antigen | XIAP |
---|---|
Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29391535), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056183T
![]() |
Novus Biologicals
NB10056183SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB10056183
![]() |
Novus Biologicals
NB10056183 |
0.05 mL |
Each for $366.00
|
|
|||||
Description
XIAP Polyclonal specifically detects XIAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
XIAP | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer | |
Whole antisera with 0.05% Sodium Azide | |
331 | |
Recombinant BIR2 domain protein fragment of human XIAP was used as immunogen. The BIR2 domain used for immunogen corresponds to amino acids 163-230 of human XIAP (Deveraux et al, 1999). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1:1000-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29391535), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat | |
API3X-linked inhibitor of apoptosis protein, apoptosis inhibitor 3, baculoviral IAP repeat-containing 4, baculoviral IAP repeat-containing protein 4, BIRC4XLP2, E3 ubiquitin-protein ligase XIAP, EC 6.3.2.-, hIAP3, hIAP-3, hILP, IAP3, IAP-3, IAP-like protein, ILP, ILP1, Inhibitor of apoptosis protein 3, MIHA, X-linked IAP, X-linked inhibitor of apoptosis | |
XIAP | |
IgG | |
Unpurified | |
The antibody recognizes epitopes in the BIR2 domain of XIAP. Therefore it can recognize full-length XIAP and XIAP cleavage fragments containing the BIR2 domain. However, XIAP cleavage fragments may be biologically unstable, and therefore cleavage fragments may be difficult to detect. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title