Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XIAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.00 - $546.00
Specifications
Antigen | XIAP |
---|---|
Concentration | 0.2 mg/mL |
Dilution | Western Blot |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018260
![]() |
Novus Biologicals
NB6008720.025MG |
0.025 mg |
Each for $243.00
|
|
|||||
NB60087201
![]() |
Novus Biologicals
NB6008720.1MG |
0.1 mg |
Each for $546.00
|
|
|||||
Description
XIAP Polyclonal specifically detects XIAP in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
XIAP | |
Western Blot | |
Polyclonal | |
Rabbit | |
Human, Rat, Mouse | |
P98170 | |
331 | |
Synthetic peptide surrounding amino acid 230 of human XIAP (ID# BV-94) | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
57 kDa |
0.2 mg/mL | |
Western Blot | |
Unconjugated | |
Apoptosis, Cancer | |
PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal | |
API3X-linked inhibitor of apoptosis protein, apoptosis inhibitor 3, baculoviral IAP repeat-containing 4, baculoviral IAP repeat-containing protein 4, BIRC4XLP2, E3 ubiquitin-protein ligase XIAP, EC 6.3.2.-, hIAP3, hIAP-3, hILP, IAP3, IAP-3, IAP-like protein, ILP, ILP1, Inhibitor of apoptosis protein 3, MIHA, X-linked IAP, X-linked inhibitor of apoptosis | |
XIAP | |
IgG | |
Affinity Purified | |
This detects the 53 kDa human XIAP protein. It does not cross-react with other apoptosis inhibitors. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title