Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XIAP Monoclonal Antibody (1020CT7.2.2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537682
Description
XIAP Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
ATP synthase is extremely conserved through evolution and can be found in plants, fungi, bacteria, and animals. The ATP synthase enzyme is a transmembrane protein responsible for driving the reversible reaction from ADP+ phosphate to ATP. This reaction is accomplished by a flux of protons across the membrane as a result of electron transfer. The ATP synthase protein has two main sections; the F1 ATP-ase (soluble) and the F0 ATP-ase (membrane embedded). The F1 section consists of the alpha, beta, gamma, delta, and epsilon subunits. While the F0 consists of a, b, and c subunits.
Specifications
XIAP | |
Monoclonal | |
0.50 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P98170 | |
XIAP | |
recombinant protein human XIAP. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1020CT7.2.2 | |
Unconjugated | |
XIAP | |
1110015C02Rik; Aipa; Api3; apoptosis inhibitor protein 3; baculoviral IAP repeat-containing 4; baculoviral IAP repeat-containing protein 4; BIR4; Birc4; E3 ubiquitin-protein ligase XIAP; hIAP3; hIAP-3; HILP; IAP homolog A; IAP3; IAP-3; IAP-like protein; ILP; ILP1; ILP-1; Inhibitor of apoptosis protein 3; MIHA; riap3; RIAP-3; RING-type E3 ubiquitin transferase XIAP; RP1-315G1.5; XIAP; X-linked IAP; X-linked inhibitor of apoptosis; X-linked inhibitor of apoptosis protein; X-linked inhibitor of apoptosis, E3 ubiquitin protein ligase; XLP2 | |
Mouse | |
Protein G | |
RUO | |
331 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction