Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XIAP Monoclonal Antibody (4C12G10)
GREENER_CHOICE

Catalog No. FERMA548373
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548373 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548373 Supplier Invitrogen™ Supplier No. MA548373
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XIAP Monoclonal Antibody for Western Blot, Flow, ELISA

ATP synthase is extremely conserved through evolution and can be found in plants, fungi, bacteria, and animals. The ATP synthase enzyme is a transmembrane protein responsible for driving the reversible reaction from ADP+ phosphate to ATP. This reaction is accomplished by a flux of protons across the membrane as a result of electron transfer. The ATP synthase protein has two main sections; the F1 ATP-ase (soluble) and the F0 ATP-ase (membrane embedded). The F1 section consists of the alpha, beta, gamma, delta, and epsilon subunits. While the F0 consists of a, b, and c subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XIAP
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4C12G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene XIAP
Gene Accession No. P98170
Gene Alias 1110015C02Rik; Aipa; Api3; apoptosis inhibitor protein 3; baculoviral IAP repeat-containing 4; baculoviral IAP repeat-containing protein 4; BIR4; Birc4; E3 ubiquitin-protein ligase XIAP; hIAP3; hIAP-3; HILP; IAP homolog A; IAP3; IAP-3; IAP-like protein; ILP; ILP1; ILP-1; Inhibitor of apoptosis protein 3; MIHA; riap3; RIAP-3; RING-type E3 ubiquitin transferase XIAP; RP1-315G1.5; XIAP; X-linked IAP; X-linked inhibitor of apoptosis; X-linked inhibitor of apoptosis protein; X-linked inhibitor of apoptosis, E3 ubiquitin protein ligase; XLP2
Gene Symbols XIAP
Host Species Mouse
Immunogen Purified recombinant fragment of human XIAP (AA: 1-268) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 331
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.