Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XIAP Mouse anti-Human, Mouse, Rat, Clone: 2A1A12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | XIAP |
---|---|
Clone | 2A1A12 |
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7199
![]() ![]() |
Proteintech Group Inc
668001IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7.Specifications
XIAP | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
API3, BIRC4, hIAP 3, hIAP3, hILP, IAP 3, IAP like protein, IAP3, ILP, ILP1, MIHA, X linked IAP, XIAP, XLP2 | |
XIAP | |
IgG1 | |
Protein G | |
Antibody |
2A1A12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P98170, Q60989, Q9R0I6 | |
11798, 331, 63879 | |
XIAP Fusion Protein Ag18088 | |
Primary | |
-20°C | |
XIAP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title