Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XIAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA513116
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513116 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513116 Supplier Invitrogen™ Supplier No. PA513116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XIAP Polyclonal Antibody for Western Blot, IHC (P)

ATP synthase is extremely conserved through evolution and can be found in plants, fungi, bacteria, and animals. The ATP synthase enzyme is a transmembrane protein responsible for driving the reversible reaction from ADP+ phosphate to ATP. This reaction is accomplished by a flux of protons across the membrane as a result of electron transfer. The ATP synthase protein has two main sections; the F1 ATP-ase (soluble) and the F0 ATP-ase (membrane embedded). The F1 section consists of the alpha, beta, gamma, delta, and epsilon subunits. While the F0 consists of a, b, and c subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XIAP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene XIAP
Gene Accession No. P98170, Q60989, Q9R0I6
Gene Alias 1110015C02Rik; Aipa; Api3; apoptosis inhibitor protein 3; baculoviral IAP repeat-containing 4; baculoviral IAP repeat-containing protein 4; BIR4; Birc4; E3 ubiquitin-protein ligase XIAP; hIAP3; hIAP-3; HILP; IAP homolog A; IAP3; IAP-3; IAP-like protein; ILP; ILP1; ILP-1; Inhibitor of apoptosis protein 3; MIHA; riap3; RIAP-3; RING-type E3 ubiquitin transferase XIAP; RP1-315G1.5; XIAP; X-linked IAP; X-linked inhibitor of apoptosis; X-linked inhibitor of apoptosis protein; X-linked inhibitor of apoptosis, E3 ubiquitin protein ligase; XLP2
Gene Symbols XIAP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human XIAP (BIRC4).
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11798, 331, 63879
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.