Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XIRP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548605
Description
XIRP1 Polyclonal Antibody for Western Blot, IHC (P)
Protects actin filaments from depolymerization.
Specifications
XIRP1 | |
Polyclonal | |
Unconjugated | |
XIRP1 | |
AI415219; cardiomyopathy associated 1; cardiomyopathy-associated protein 1; CMYA1; LOC100910104; mXin alpha; Xin; xin actin binding repeat containing 1; xin actin-binding repeat containing 1; xin actin-binding repeat-containing protein 1; xin actin-binding repeat-containing protein 1-like; Xin alpha; XIRP1 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
165904 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q702N8 | |
XIRP1 | |
KLH conjugated synthetic peptide between 1338-1367 amino acids from the C-terminal region of human XIRP1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction