Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ XIRP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553805
Description
Immunogen sequence: RVSTEVAQLK EQTLARLLDI EEAVHKALSS MSSLQPEASA RGHFQGPPKD HSAHKISVTV SSSARPSGSG QEVGGQTAVK NQAKVECHTE AQSQVKIRNH T Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 27%.
Protects actin filaments from depolymerization.
Specifications
XIRP1 | |
Polyclonal | |
Unconjugated | |
XIRP1 | |
AI415219; cardiomyopathy associated 1; cardiomyopathy-associated protein 1; CMYA1; LOC100910104; mXin alpha; Xin; xin actin binding repeat containing 1; xin actin-binding repeat containing 1; xin actin-binding repeat-containing protein 1; xin actin-binding repeat-containing protein 1-like; Xin alpha; XIRP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
165904 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q702N8 | |
XIRP1 | |
Recombinant protein corresponding to Human XIRP1. Recombinant protein control fragment (Product #RP-92429). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction