Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XIRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557072
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557072 Supplier Invitrogen™ Supplier No. PA557072
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HAPPTYEDVI AGHILDISDS PKEVRKNFQK TWQESGRVFK GLGYATADAS ATEMRTTFQE ESAFISEAAA PRQGNMYTLS KDSLSNGVP Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen XIRP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XIRP2
Gene Accession No. A4UGR9
Gene Alias 2310003D02Rik; 2310008C07Rik; A530024P18Rik; AI452089; Beta-xin; cardiomyopathy associated 3; cardiomyopathy-associated protein 3; CMYA3; Gm352; mXinbeta; Myogenic MEF2-activated Xin-related protein; myomaxin; Xeplin; xin actin binding repeat containing 2; xin actin-binding repeat containing 2; Xin actin-binding repeat containing 2 short splice form; xin actin-binding repeat-containing protein 2; xin repeat protein 2; Xin2; xin-beta; Xirp2
Gene Symbols XIRP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human XIRP2. Recombinant protein control fragment (Product #RP-96176).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129446
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.