Learn More
Description
XK columns are constructed of robust borosilicate glass, which exhibits excellent chemical resistance and is compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules. The clear tubes allow for visual inspection of the media bed and are available in a broad range of column dimensions for use with many different purification needs.
Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations.
- Plunger design to ensure a homogeneous buffer distribution and reproducible packing
- Adapter QuickLock mechanism that facilitates column handling and cleaning
- Fittings for direct connection to chromatographiy systems
- Cooling jacket for sensitive purifications
- Extensive range of spare parts available, facilitating maintenance
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | XK Chromatography Columns (Empty) |
| Bed Volume | Bed height (one adapter): 530 to 650mm Bed height (two adapters): 385 to 650mm |
| Diameter (Metric) Inner | 26 mm |
| Height (Metric) | 700 mm |
| Flow Rate | Inlet/Outlet (i.d.): 0.75mm |
| Material | Borosilicate glass |
| Max. Pressure | 5 bar |
| Particle Size | 10μm pore size |
| Support Type | Polypropylene (reinforced glass fiber)/Polyamide Net |
| Column Type | Empty Chromatography Column |
| Show More |
Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol.
Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
