Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XKR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551917
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551917 Supplier Invitrogen™ Supplier No. PA551917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TIRNYHKWLK NLKQEKEETQ VSITKRNTML EREIAFSIRD NFMQQKAFK Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 31%.

XKRX (MIM 300684) and XKR3 are homologs of the Kell blood group precursor XK (MIM 314850), which is a putative membrane transporter and a component of the XK/Kell complex of the Kell blood group system (Calenda et al., 2006 [PubMed 16431037]).[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen XKR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XKR3
Gene Accession No. Q5GH77
Gene Alias expressed in testis; X Kell blood group precursor-related family, member 3; x Kell blood group-related 3; XK related 3; XK, Kell blood group complex subunit-related family, member 3; XKR3; XK-related protein 3; X-linked Kx blood group related 3; XRG3; XTES
Gene Symbols XKR3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human XKR3. Recombinant protein control fragment (Product #RP-91537).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150165
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.