Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XKR5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571286
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571286 Supplier Invitrogen™ Supplier No. PA571286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XKR5 Polyclonal Antibody for Western Blot

XKR5 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XKR5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene XKR5
Gene Accession No. Q6UX68
Gene Alias HARL2754; UNQ2754; UNQ2754/PRO6493; X Kell blood group precursor-related family, member 5; XK related 5; XK, Kell blood group complex subunit-related family, member 5; XKR5; XK-related protein 5; XK-related protein 5a; X-linked Kx blood group related 5; XRG5; XRG5A; XRG5BM
Gene Symbols XKR5
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human XKR5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389610
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.