Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XKR6 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306015JF646
Description
XKR6 Polyclonal specifically detects XKR6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
XKR6 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C8orf5, C8orf7, FLJ31557, Transmembrane protein C8orf5, X Kell blood group precursor-related family, member 6, XK, Kell blood group complex subunit-related family, member 6, XK-related protein 6, XRG6 | |
Produced in rabbits immunized with E. coli-derived Human XKR6 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
286046 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction